3-Fluoro-4-(methoxycarbonyl)benzoic acid 95%

Supplier: Ambeed
Warning

A225289-100MG A225289-1G A225289-250MG A225289-5G
76647-580EA 21 USD
76647-580 76647-582 76647-584 76647-586
3-Fluoro-4-(methoxycarbonyl)benzoic acid 95%
3-Fluoro-4-(methoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₄
MDL Number: MFCD20639684
CAS Number: 161796-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR