5-Bromo-2-chloro-3-(trifluoromethyl)thiophene 97%
Supplier: Ambeed
|
Warning
|
A223589-1G
A223589-250MG
76647-566EA
1032.26
USD
76647-566
76647-568
5-Bromo-2-chloro-3-(trifluoromethyl)thiophene 97%
5-Bromo-2-chloro-3-(trifluoromethyl)thiophene
Precaución: for Research Use Only
Formula:
C₅HBrClF₃S MW: 265.4787 g/mol |
CAS Number:
2168435-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...