7-Isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-1-oxo- 95%

Supplier: Ambeed
Warning

A222063-1G A222063-250MG
76647-562EA 534.7 USD
76647-562 76647-564
7-Isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-1-oxo- 95%
7-Isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-1-oxo-

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.1834 g/mol
MDL Number: MFCD19683459
CAS Number: 1343932-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR