5-Phenyl-1H-pyrazol-3-ol 97%

Supplier: Ambeed
Warning

A221473-5G A221473-1G
76647-560EA 180.36 USD
76647-560 77748-357
5-Phenyl-1H-pyrazol-3-ol 97%
5-Phenyl-1H-pyrazol-3-ol

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.1726 g/mol
MDL Number: MFCD02685648
CAS Number: 145091-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR