5-Bromo-3-(piperidin-1-yl)pyridin-2-amine 95%

Supplier: Ambeed
Warning

A220547-1G A220547-250MG
76647-554EA 116.7 USD
76647-554 76647-556
5-Bromo-3-(piperidin-1-yl)pyridin-2-amine 95%
5-Bromo-3-(piperidin-1-yl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrN₃
MW: 256.1423 g/mol
MDL Number: MFCD30528819
CAS Number: 1335058-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR