2-Methoxy-1H-indole 98%

Supplier: Ambeed
Warning

A219063-100MG A219063-1G A219063-250MG
76647-548EA 137.92 USD
76647-548 76647-550 76647-552
2-Methoxy-1H-indole 98%
2-Methoxy-1H-indole

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.1739 g/mol
CAS Number: 1184915-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR