(2-Bromo-4,5-dimethoxyphenyl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Bromo-4,5-dimethoxyphenyl)acetonitrile

A218845-1G A218845-250MG A218845-25G A218845-10G
76647-542EA 14.86 USD
76647-542 76647-544 76647-546 77615-838
(2-Bromo-4,5-dimethoxyphenyl)acetonitrile 97%
(2-Bromo-4,5-dimethoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO₂
MW: 256.1000 g/mol
MDL Number: MFCD00016385
CAS Number: 51655-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR