3-Fluorobicyclo[1.1.1]pentan-1-amine hydrochloride 98%

Supplier: Ambeed
Warning

A218746-100MG A218746-1G
76647-540EA 159.14 USD
76647-540 77550-420
3-Fluorobicyclo[1.1.1]pentan-1-amine hydrochloride 98%
3-Fluorobicyclo[1.1.1]pentan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈FN·ClH
MW: 137.583 g/mol
MDL Number: MFCD28963742
CAS Number: 1826900-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR