Benzyl (R)-2-(cyanomethyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A1339098-100MG A1339098-250MG
76657-972EA 80.63 USD
76657-972 76657-974
Benzyl (R)-2-(cyanomethyl)piperazine-1-carboxylate 95%
Benzyl (R)-2-(cyanomethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇N₃O₂
MW: 259.3037 g/mol
MDL Number: MFCD31925385
CAS Number: 2306255-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR