3-Chloroisoxazolo[4,5-b]pyridine 95%

Supplier: Ambeed
Warning

A212783-100MG A212783-250MG A212783-1G
76647-528EA 101 USD
76647-528 76647-530 77748-324
3-Chloroisoxazolo[4,5-b]pyridine 95%
3-Chloroisoxazolo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClN₂O
MDL Number: MFCD28515540
CAS Number: 1784989-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR