5-Methoxyquinazolin-4(3H)-one 97%

Supplier: Ambeed
Warning

A209838-100MG A209838-5G
76647-514EA 78.51 USD
76647-514 76647-516
5-Methoxyquinazolin-4(3H)-one 97%
5-Methoxyquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.1720 g/mol
MDL Number: MFCD13191850
CAS Number: 135106-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR