3-Methyl-5-(thiophen-2-yl)-1H-pyrrole-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A207377-100MG
A207377-250MG
76647-498EA
152.77
USD
76647-498
76647-500
3-Methyl-5-(thiophen-2-yl)-1H-pyrrole-2-carboxylic acid 97%
3-Methyl-5-(thiophen-2-yl)-1H-pyrrole-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₂S MW: 207.2500 g/mol |
CAS Number:
2090492-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...