tert-Butyl (R)-3-(cyanomethyl)piperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A1339097-100MG A1339097-250MG A1339097-1G
76657-968EA 143.23 USD
76657-968 76657-970 77621-586
tert-Butyl (R)-3-(cyanomethyl)piperazine-1-carboxylate 98%
tert-Butyl (R)-3-(cyanomethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉N₃O₂
MW: 225.2875 g/mol
MDL Number: MFCD32659992
CAS Number: 2306248-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR