5-(3-Hydroxyprop-1-en-1-yl)-2-methoxyphenol 95%

Supplier: Ambeed
Warning

A205531-1G A205531-250MG
76647-488EA 128 USD
76647-488 76647-490
5-(3-Hydroxyprop-1-en-1-yl)-2-methoxyphenol 95%

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
CAS Number: 374706-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR