3-Fluoro-2-methoxy-5-pyridineboronic acid 98%

Supplier: Ambeed
Warning

A203471-100MG A203471-1G A203471-250MG A203471-5G
76647-478EA 12 USD
76647-478 76647-480 76647-482 76647-484
3-Fluoro-2-methoxy-5-pyridineboronic acid 98%
3-Fluoro-2-methoxy-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₃
MDL Number: MFCD04112520
CAS Number: 856250-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR