Methyl 4-bromo-2-formylbenzoate 95%

Supplier: Ambeed
Warning

A202108-100MG A202108-250MG
76647-470EA 93.36 USD
76647-470 76647-472
Methyl 4-bromo-2-formylbenzoate 95%
Methyl 4-bromo-2-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
MW: 243.054 g/mol
MDL Number: MFCD16036574
CAS Number: 1260795-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR