1-Deoxynojirimycin hydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5S)-2-(Hydroxymethyl)piperidine-3,4,5-triol hydrochloride

A198597-100MG A198597-1MG A198597-250MG
76647-452EA 78.51 USD
76647-452 76647-454 76647-456
1-Deoxynojirimycin hydrochloride 96%
1-Deoxynojirimycin hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO₄·HCl
MW: 199.64 g/mol
MDL Number: MFCD00133247
CAS Number: 73285-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR