3,4-Dibromobenzaldehyde 97%

Supplier: Ambeed
Warning

A198153-10G A198153-1G A198153-250MG A198153-25G A198153-5G
76647-442EA 73 USD
76647-442 76647-444 76647-446 76647-448 77615-680
3,4-Dibromobenzaldehyde 97%
3,4-Dibromobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Br₂O
MDL Number: MFCD00236033
CAS Number: 74003-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR