Polyethylene glycol 400 Mn 380-420 ∼ Mn 400

Supplier: Ambeed
Warning

Synonyms: PEG 400

A197578-1000G A197578-100G A197578-25G A197578-500G A197578-5G
76647-432EA 19.1 USD
76647-432 76647-434 76647-436 76647-438 76647-440
Polyethylene glycol 400 Mn 380-420 ∼ Mn 400
Polyethylene glycol 400

Precaución: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MW: 400 g/mol
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR