tert-Butyl 3-(4-nitrophenyl)azetidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A196563-100MG A196563-250MG
76647-422EA 106.09 USD
76647-422 76647-424
tert-Butyl 3-(4-nitrophenyl)azetidine-1-carboxylate 97%
tert-Butyl 3-(4-nitrophenyl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₄
MW: 278.3037 g/mol
MDL Number: MFCD20528755
CAS Number: 883901-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR