3-Bromo-1H-pyrazolo[4,3-b]pyridine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A195686-100MG A195686-1G A195686-250MG
76647-414EA 159.14 USD
76647-414 76647-416 76647-418
3-Bromo-1H-pyrazolo[4,3-b]pyridine-5-carbonitrile 95%
3-Bromo-1H-pyrazolo[4,3-b]pyridine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrN₄
MW: 223.0295 g/mol
MDL Number: MFCD30528764
CAS Number: 2091782-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR