N-Carbobenzoxy-L-leucine 97% ~3% EA

Supplier: Ambeed
Warning

Synonyms: N-Cbz-L-leucine, Z-Leu-OH

A195117-1000G A195117-100G A195117-25G A195117-500G A195117-5G
76647-402EA 624 USD
76647-402 76647-404 76647-406 76647-408 76647-410 76647-412
N-Carbobenzoxy-L-leucine 97% ~3% EA
N-Carbobenzoxy-L-leucine

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD00026494
CAS Number: 2018-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR