(S)-Piperidin-3-ylmethanol hydrochloride 97%

Supplier: Ambeed
Warning

A190231-10G A190231-1G A190231-250MG A190231-5G
76647-384EA 131 USD
76647-384 76647-386 76647-388 76647-390
(S)-Piperidin-3-ylmethanol hydrochloride 97%
(S)-Piperidin-3-ylmethanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO·ClH
MDL Number: MFCD14155681
CAS Number: 1125551-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR