2-Bromo-3-ethylpyridine 95%

Supplier: Ambeed
Warning

A188535-250MG A188535-1G
76647-374EA 180.36 USD
76647-374 77567-756
2-Bromo-3-ethylpyridine 95%
2-Bromo-3-ethylpyridine

Precaución: For research use only.

Formula: C₇H₈BrN
MW: 186.0491 g/mol
MDL Number: MFCD22394192
CAS Number: 142337-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR