2-Cyano-5-methylpyridine 1-oxide 95%

Supplier: Ambeed
Danger

A186474-1G A186474-250MG A186474-5G
76647-360EA 58.35 USD
76647-360 76647-362 76647-364
2-Cyano-5-methylpyridine 1-oxide 95%
2-Cyano-5-methylpyridine 1-oxide

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.1353 g/mol
MDL Number: MFCD27945564
CAS Number: 159727-87-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR