2-Amino-3,3,3-trifluoropropan-1-ol hydrochloride 95%

Supplier: Ambeed
Warning

A186225-100MG A186225-1G A186225-250MG A186225-5G
76647-356EA 102.91 USD
76647-356 76647-358 77615-574 77748-207
2-Amino-3,3,3-trifluoropropan-1-ol hydrochloride 95%
2-Amino-3,3,3-trifluoropropan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₃H₇ClF₃NO
MW: 165.5420 g/mol
MDL Number: MFCD10697731
CAS Number: 1196154-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR