CPI-444 98%

Supplier: Ambeed
Warning

Synonyms: V81444, Ciforadenant, (S)-7-(5-methylfuran-2-yl)-3-((6-(((tetrahydrofuran-3-yl)oxy)methyl)pyridin-2-yl)methyl)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-5-amine

A186100-10MG A186100-1MG A186100-25MG A186100-50MG A186100-5MG
76647-340EA 35 USD
76647-340 76647-342 76647-344 76647-346 76647-348
CPI-444 98%
CPI-444

Precaución: for Research Use Only

Formula: C₂₀H₂₁N₇O₃
MW: 407.43 g/mol
MDL Number: MFCD31540410
CAS Number: 1202402-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR