(R)-(+)-Citronellic acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3,7-Dimethyloct-6-enoic acid, (3R)-3,7-Dimethyloct-6-enoic acid

A185755-100MG A185755-10G A185755-1G A185755-250MG A185755-5G
76647-330EA 37.14 USD
76647-330 76647-332 76647-334 76647-336 76647-338
(R)-(+)-Citronellic acid 98%
(R)-(+)-Citronellic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MW: 170.249 g/mol
MDL Number: MFCD00075195
CAS Number: 18951-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR