4-Hydroxy-8-methoxyquinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A184563-100MG A184563-1G A184563-250MG
76647-316EA 43 USD
76647-316 76647-318 76647-320
4-Hydroxy-8-methoxyquinolin-2(1H)-one 97%
4-Hydroxy-8-methoxyquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MDL Number: MFCD02183249
CAS Number: 7224-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR