5,7-Dimethyl-N-(trans-4-(pentyloxy)cyclohexyl)pyrazolo[1,5-a]pyrimidine-3-carboxamide 99%

Supplier: Ambeed
Warning

Synonyms: Gcase activator 1

A181720-10MG A181720-1MG A181720-25MG A181720-50MG A181720-5MG A181720-100MG
76647-296EA 59.42 USD
76647-296 76647-298 76647-300 76647-302 76647-304 77567-666
5,7-Dimethyl-N-(trans-4-(pentyloxy)cyclohexyl)pyrazolo[1,5-a]pyrimidine-3-carboxamide 99%
5,7-Dimethyl-N-(trans-4-(pentyloxy)cyclohexyl)pyrazolo[1,5-a]pyrimidine-3-carboxamide

Precaución: For research use only.

Formula: C₂₀H₃₀N₄O₂
MW: 358.48 g/mol
MDL Number: MFCD31618016
CAS Number: 1919820-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR