1-Bromo-9,9'-spirobi[9H-fluorene] 99%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-9,9'-spirobifluorene

A181701-1G A181701-5G A181701-25G
76647-292EA 32.89 USD
76647-292 76647-294 77622-258
1-Bromo-9,9'-spirobi[9H-fluorene] 99%
1-Bromo-9,9'-spirobi[9H-fluorene]

Precaución: for Research Use Only

Formula: C₂₅H₁₅Br
MW: 395.30 g/mol
MDL Number: MFCD31556229
CAS Number: 1450933-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR