6-Chloro-1H-indazole-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A180616-100MG A180616-250MG
76647-280EA 94.43 USD
76647-280 76647-282
6-Chloro-1H-indazole-5-carboxylic acid 98%
6-Chloro-1H-indazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MW: 196.5905 g/mol
MDL Number: MFCD31537127
CAS Number: 1890961-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR