tert-Butyl (2-chlorothiazol-4-yl)carbamate 95%

Supplier: Ambeed
Warning

A178414-100MG A178414-1G A178414-250MG
76647-264EA 31.83 USD
76647-264 76647-266 76647-268
tert-Butyl (2-chlorothiazol-4-yl)carbamate 95%
tert-Butyl (2-chlorothiazol-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O₂S
MW: 234.7031 g/mol
MDL Number: MFCD30188981
CAS Number: 1258934-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR