4-(5-(2,6-Dimethoxyphenyl)-3-(methoxycarbonyl)-1H-pyrazol-1-yl)-3-isopropylbenzoic acid 95%

Supplier: Ambeed
Warning

A175419-250MG A175419-1G
76647-248EA 50.93 USD
76647-248 77748-145
4-(5-(2,6-Dimethoxyphenyl)-3-(methoxycarbonyl)-1H-pyrazol-1-yl)-3-isopropylbenzoic acid 95%
4-(5-(2,6-Dimethoxyphenyl)-3-(methoxycarbonyl)-1H-pyrazol-1-yl)-3-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₄N₂O₆
MW: 424.4465 g/mol
MDL Number: MFCD31698187
CAS Number: 184163-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR