tert-Butyl 9-bromononanoate 97%

Supplier: Ambeed
Warning

A172678-100MG A172678-250MG A172678-1G A172678-5G
76647-238EA 34 USD
76647-238 76647-240 77567-538 77783-352
tert-Butyl 9-bromononanoate 97%
tert-Butyl 9-bromononanoate

Precaución: For research use only.

Formula: C₁₃H₂₅BrO₂
MDL Number: MFCD31728663
CAS Number: 135982-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR