(S)-1-(4-Fluorophenyl)-1-(2-(4-(6-(1-methyl-1H-pyrazol-4-yl)pyrrolo[2,1-f][1,2,4]triazin-4-yl)piperazin-1-yl)pyrimidin-5-yl)ethan-1-amine 98%

Supplier: Ambeed
Warning

Synonyms: Avapritinib

A172367-5MG A172367-1MG A172367-100MG A172367-50MG
76647-236EA 51.99 USD
76647-236 77593-082 77615-444 77615-446
(S)-1-(4-Fluorophenyl)-1-(2-(4-(6-(1-methyl-1H-pyrazol-4-yl)pyrrolo[2,1-f][1,2,4]triazin-4-yl)piperazin-1-yl)pyrimidin-5-yl)ethan-1-amine 98%
(S)-1-(4-Fluorophenyl)-1-(2-(4-(6-(1-methyl-1H-pyrazol-4-yl)pyrrolo[2,1-f][1,2,4]triazin-4-yl)piperazin-1-yl)pyrimidin-5-yl)ethan-1-amine

Precaución: for Research Use Only

Formula: C₂₆H₂₇FN₁₀
MW: 498.56 g/mol
MDL Number: MFCD31544325
CAS Number: 1703793-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR