Polyethylene glycol 6,000 ∼ Mn 6000
Supplier: Ambeed
|
Warning
|
Synonyms:
PEG 6000
, PEG 6,000
A172254-100G
A172254-1000G
A172254-25G
A172254-500G
76647-230EA
9.27
USD
76647-230
76647-228
76647-224
76647-226
Polyethylene glycol 6,000 ∼ Mn 6000
Polyethylene glycol 6,000
Precaución: for Research Use Only
Formula:
H(OCH₂CH₂)nOH MW: 6000 g/mol |
MDL Number:
MFCD01779614 CAS Number: 25322-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...