2-(Chloromethyl)-8-methylquinazolin-4(3H)-one 95%

Supplier: Ambeed
Danger

A171805-100MG A171805-1G A171805-250MG
76647-218EA 23 USD
76647-218 76647-220 76647-222
2-(Chloromethyl)-8-methylquinazolin-4(3H)-one 95%
2-(Chloromethyl)-8-methylquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O
MW: 208.6443 g/mol
MDL Number: MFCD07324008
CAS Number: 848369-52-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR