Ethyl 6-chloro-3-nitropicolinate 95%

Supplier: Ambeed
Warning

A167842-250MG A167842-100MG A167842-1G
76647-210EA 32 USD
76647-210 77748-116 77748-117
Ethyl 6-chloro-3-nitropicolinate 95%
Ethyl 6-chloro-3-nitropicolinate

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O₄
MDL Number: MFCD28757779
CAS Number: 1805648-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR