Ethyl 7-fluoro-4-methoxy-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

A164474-100MG A164474-1G A164474-250MG
76647-182EA 83.82 USD
76647-182 76647-184 76647-186
Ethyl 7-fluoro-4-methoxy-1H-indole-2-carboxylate 97%
Ethyl 7-fluoro-4-methoxy-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂FNO₃
MW: 237.2270 g/mol
MDL Number: MFCD18251689
CAS Number: 1197943-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR