2-(5-Bromo-1H-indol-3-yl)-1-ethanol 97%

Supplier: Ambeed
Warning

A164364-100MG A164364-10G A164364-1G A164364-250MG A164364-5G
76647-172EA 17 USD
76647-172 76647-174 76647-176 76647-178 76647-180
2-(5-Bromo-1H-indol-3-yl)-1-ethanol 97%
2-(5-Bromo-1H-indol-3-yl)-1-ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MDL Number: MFCD00130167
CAS Number: 32774-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR