4-Methylpyridin-3-ol 95%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-4-methylpyridine

A163502-1G A163502-25G A163502-5G
76647-164EA 18.04 USD
76647-164 76647-166 76647-168
4-Methylpyridin-3-ol 95%
4-Methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.1259 g/mol
MDL Number: MFCD04114227
CAS Number: 1121-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR