1-(4-Amino-3-chlorophenyl)cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

A161154-100MG A161154-1G A161154-250MG
76647-148EA 47.75 USD
76647-148 76647-150 76647-152
1-(4-Amino-3-chlorophenyl)cyclopropanecarboxylic acid 97%
1-(4-Amino-3-chlorophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClNO₂
MW: 211.6449 g/mol
CAS Number: 1314738-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR