4-(Cyclopropylethynyl)benzoic acid 95%

Supplier: Ambeed
Warning

A160802-100MG A160802-250MG
76647-144EA 60.48 USD
76647-144 76647-146
4-(Cyclopropylethynyl)benzoic acid 95%
4-(Cyclopropylethynyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.2066 g/mol
MDL Number: MFCD17171061
CAS Number: 908247-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR