9-([1,1'-Biphenyl]-4-yl)-9'-phenyl-9H,9'H-3,3'-bicarbazole 95%

Supplier: Ambeed
Warning

A160765-1G A160765-250MG A160765-5G
76647-138EA 67.9 USD
76647-138 76647-140 76647-142
9-([1,1'-Biphenyl]-4-yl)-9'-phenyl-9H,9'H-3,3'-bicarbazole 95%
9-([1,1'-Biphenyl]-4-yl)-9'-phenyl-9H,9'H-3,3'-bicarbazole

Precaución: for Research Use Only

Formula: C₄₂H₂₈N₂
MW: 560.6900 g/mol
MDL Number: MFCD31735647
CAS Number: 1454567-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR