3,3-Dimethylindolin-7-amine hydrochloride 90+%

Supplier: Ambeed
Warning

A160058-100MG A160058-1G A160058-250MG
76647-132EA 90.18 USD
76647-132 76647-134 76647-136
3,3-Dimethylindolin-7-amine hydrochloride 90+%
3,3-Dimethylindolin-7-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₅ClN₂
MW: 198.6925 g/mol
MDL Number: MFCD31715097
CAS Number: 2682112-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR