3-(3-(3-Hydroxyphenyl)prop-2-yn-1-yl)-1,7-dimethyl-8-(methylsulfonyl)-3,7-dihydro-1H-purine-2,6-dione 97%

Supplier: Ambeed
Warning

A157358-1MG A157358-5MG
76647-112EA 182 USD
76647-112 76647-114
3-(3-(3-Hydroxyphenyl)prop-2-yn-1-yl)-1,7-dimethyl-8-(methylsulfonyl)-3,7-dihydro-1H-purine-2,6-dione 97%
3-(3-(3-Hydroxyphenyl)prop-2-yn-1-yl)-1,7-dimethyl-8-(methylsulfonyl)-3,7-dihydro-1H-purine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₄O₅S
MW: 388.4 g/mol
CAS Number: 2093393-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR