3-Bromo-5,7-dichloro-1H-pyrazolo[4,3-d]pyrimidine 95%

Supplier: Ambeed
Danger

A156613-100MG A156613-250MG A156613-50MG A156613-1G
76647-106EA 145.35 USD
76647-106 76647-108 76647-110 77567-278
3-Bromo-5,7-dichloro-1H-pyrazolo[4,3-d]pyrimidine 95%
3-Bromo-5,7-dichloro-1H-pyrazolo[4,3-d]pyrimidine

Precaución: For research use only.

Formula: C₅HBrCl₂N₄
MW: 267.8982 g/mol
MDL Number: MFCD29761463
CAS Number: 1934543-12-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR