6-Bromo-5-methylquinoxaline 95%

Supplier: Ambeed
Warning

A155897-100MG A155897-250MG
76647-092EA 53 USD
76647-092 76647-094
6-Bromo-5-methylquinoxaline 95%
6-Bromo-5-methylquinoxaline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD31699512
CAS Number: 2386229-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR