Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II) 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Ru(OAc)2(BINAP), Ru(OAc)2[(S)-binap]

A152145-250MG A152145-1G A152145-5G A152145-25G
76647-048EA 39.26 USD
76647-048 77597-932 77597-934 77615-234
Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II) 98%
Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)

Precaución: for Research Use Only

Formula: C₄₈H₃₈O₄P₂Ru
MW: 841.83 g/mol
MDL Number: MFCD09753020
CAS Number: 261948-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR